Learn More
SAI Infusion Technologies Stainless Steel Catheter Connectors - 21G, 1cm
SDP
Catalog No. NC0737981
Encompass
SAI's catheter connectors ensure a safe and effective fit when connecting fluid lines for seamless fluid flow. Burr-free stainless steel prevents damage to the catheter during insertion and removal. 21G, 1cm, 100/pk. To plug a catheter, use solid stainless steel catheter pins (CP-21). Product is for laboratory animal research only.
Catalog No. NC0737981 Supplier SAI Infusion Technologies Supplier No. CC21
SAI's catheter connectors ensure a safe and effective fit when connecting fluid lines for seamless fluid flow. Burr-free stainless steel prevents damage to the catheter during insertion and removal. 21G, 1cm, 100/pk. To plug a catheter, use solid stainless steel catheter pins (CP-21). Product is for laboratory animal research only.