Learn More
Medchemexpress LLC 21-hydroxyhenicosanoic acid | 89160-04-3 | MFCD02259051 | 99.8% | 342.56 g/mol | C21H42O3 | 5 G

Supplier: Medchemexpress LLC HYW6147245G
21-Hydroxyhenicosanoic acid is a long-chain omega-hydroxy fatty acid provided as a research-grade ester reagent. It is supplied in high purity and is suitable for biochemical studies, organic synthesis, and analytical reference work. Available as a solid and as a DMSO solution for convenience in solution-phase experiments.
- High purity: 99.75% (analytical grade).
- CAS number: 89160-04-3 for unambiguous identification.
- Chemical formula: C21H42O3; molecular weight ~342.6 g/mol.
- Available in common research sizes including 250 mg, 1 g, and 5 g, and as a 10 mM solution in DMSO.
- Suitable for use as an analytical standard or synthetic intermediate in laboratory research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.