Learn More
Enzo Life Sciences 21-Hydroxyoligomycin A (1mg). CAS: 102042-09-1
SDP
Supplier:  Enzo Life Sciences ALX380224M001
Encompass_Preferred
Co-metabolite of nemadectin. Antitumor agent with weak antifungal and nematocidal activity. Alternative name: Nemadectin omega, Antibiotic LL-F28249 omega. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8983