Learn More
Medchemexpress LLC 21-Hydroxypregnenolone | 1164-98-3 | 98.1% | 332.48 | 1 ML
SDP
Catalog No. 5000439604
Encompass_Preferred
21-Hydroxypregnenolone is an essential intermediate in corticosterone synthesis. It is classified as an endogenous metabolite and is related to metabolic diseases and steroids. This product is intended for research and analytical applications.
  • Essential intermediate in corticosterone synthesis
  • Classified as an endogenous metabolite
  • Related to metabolic diseases and steroids
  • Suitable for research and analytical applications
Catalog No. 50-004-39604 Supplier Medchemexpress LLC Supplier No. HY11302010MM/1ML
21-Hydroxypregnenolone is an essential intermediate in corticosterone synthesis. It is classified as an endogenous metabolite and is related to metabolic diseases and steroids. This product is intended for research and analytical applications.
  • Essential intermediate in corticosterone synthesis
  • Classified as an endogenous metabolite
  • Related to metabolic diseases and steroids
  • Suitable for research and analytical applications