Learn More
Andwin Scientific 21" White Disposable Bouffant Cap, 1000/case
SDP
Catalog No. NC1844746
Encompass_Preferred
This bouffant cap features an elastic band for snug, adjustable sizing and is designed for full hair coverage. To minimize the threat of allergens in the workplace, this cap is latex free. Made of spunbound polypropylene (SPP) fabric that is conveniently lint free, this cap is a must-have addition to any foodservice or healthcare facility where cleanliness is of the utmost importance!
This item is not returnable. View return policy
Catalog No. NC1844746 Supplier Andwin Scientific Supplier No. RP110NW
This bouffant cap features an elastic band for snug, adjustable sizing and is designed for full hair coverage. To minimize the threat of allergens in the workplace, this cap is latex free. Made of spunbound polypropylene (SPP) fabric that is conveniently lint free, this cap is a must-have addition to any foodservice or healthcare facility where cleanliness is of the utmost importance!