Learn More
Ace Glass, Inc. 210 CHEMRAZ O-RING 210 CHEMRAZ O-RING
SDP
Catalog No. NC0263469
Encompass
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Chemraz 514 A is a white perfluoroelastomer with the broadest chemical resistance of any elestomeric material. Avoid acids, aldehydes, ethylene oxide and propylene oxide. Temp range -30 to 220 deg.C
Catalog No. NC0263469 Supplier Ace Glass, Inc. Supplier No. 7859530
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Chemraz 514 A is a white perfluoroelastomer with the broadest chemical resistance of any elestomeric material. Avoid acids, aldehydes, ethylene oxide and propylene oxide. Temp range -30 to 220 deg.C