Learn More
Hans Rudolph Inc Three-Way T-Shape™ Manual Directional Control Stopcock-Type™ valves
SDP
Catalog No. NC2120910
Encompass_Preferred
Three-Way T-Shape™ Manual Directional Control Stopcock-Type™ valves available in four flow bore sizes control the direction of flow from one common side port to one of the other two selective ports. Rotary actuation, clear housing, o-ring seals and removable port adapters. Reusable.
This item is not returnable. View return policy
Catalog No. NC2120910 Supplier Hans Rudolph Inc Supplier No. 112230
Three-Way T-Shape™ Manual Directional Control Stopcock-Type™ valves available in four flow bore sizes control the direction of flow from one common side port to one of the other two selective ports. Rotary actuation, clear housing, o-ring seals and removable port adapters. Reusable.