Learn More
Hach Company 2100Q IS Port Turb LED
Catalog No. 5000207659 Shop All Hach Company Products
Encompass
2100Q IS Portable Turbidimeter (LED)-0-1000 FNU-case assembly-4 AA alkaline batteries-6 sample cells-Stablcal ampule kit-silicone oil-oiling cloth-manual
Catalog No. 50-002-07659 Supplier Hach Company Supplier No. 2100QIS01
2100Q IS Portable Turbidimeter (LED)-0-1000 FNU-case assembly-4 AA alkaline batteries-6 sample cells-Stablcal ampule kit-silicone oil-oiling cloth-manual