Learn More
Ace Glass, Inc. -212 CHEMRAZ O-RING -212 CHEMRAZ O-RING
SDP
Catalog No. 509558113
Encompass
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Chemraz 514 A is a white perfluoroelastomer with the broadest chemical resistance of any elestomeric material. Avoid acids, aldehydes, ethylene oxide and propylene oxide. Temp range -30 to 220 deg.C
Catalog No. 50-955-8113 Supplier Ace Glass, Inc. Supplier No. 7859534
Manufactured by Greene, Tweed & Company and Supplied by Ace Glass Inc. Chemraz 514 A is a white perfluoroelastomer with the broadest chemical resistance of any elestomeric material. Avoid acids, aldehydes, ethylene oxide and propylene oxide. Temp range -30 to 220 deg.C