Learn More
Ace Glass, Inc. -212 KALREZ O-RING (TG)
SDP
Catalog No. 509558040
Encompass
Size 212 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.
Catalog No. 50-955-8040 Supplier Ace Glass, Inc. Supplier No. 7855634
Size 212 Kalrez o-ring. Kalrez is a perfluoroelastomer with high temperature and chemical resistance. Has all the properties and inertness of PTFE. Can be used in pharmaceutical, food or semiconductor applications.