Learn More
Ace Glass, Inc. -212 O-RING FETFE 6/PK -212 O-RING FETFE 6/PK
SDP
Catalog No. 509558058
Encompass_Preferred
Size 212 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.
Catalog No. 50-955-8058 Supplier Ace Glass, Inc. Supplier No. 7855734
Size 212 FETFE o-ring, Pack/6. An ACE exclusive compound which is a fluoroelastomer with TFE additives. It has good compression set along w/ excellent temperature and chemical compatibility. Generally black or white. USP Class VI material.