Learn More
Waters Corp Encapsulated O-Ring, AS568-214, 0.984 in. ID, 0.139 in. CS
Catalog No. 50819099 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This encapsulated O-ring is designed for use in critical applications requiring reliable sealing. It conforms to AS568-214 sizing standards, offering precise fit and performance.
  • Conforms to AS568-214 sizing standards
  • Inside diameter of 0.984 in.
  • Cross-section of 0.139 in.
  • Suitable for ion block assembly in SQ Detector 2
Catalog No. 50-819-099 Supplier Waters Corp Supplier No. 700001897
This encapsulated O-ring is designed for use in critical applications requiring reliable sealing. It conforms to AS568-214 sizing standards, offering precise fit and performance.
  • Conforms to AS568-214 sizing standards
  • Inside diameter of 0.984 in.
  • Cross-section of 0.139 in.
  • Suitable for ion block assembly in SQ Detector 2