Learn More
BacterioScan™ 216Dx Microbial Multicuvettes
Catalog No. 22011105
Change view
Click to view available options
Type:
Filtration Multicuvette
Multicell Cuvette
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Sterility
22-011-105 Filtration Multicuvette Sterile
22-011-120 Multicell Cuvette Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-011-105 Supplier BacterioScan™ Supplier No. 4DXSLB5064
Multicuvettes: Filtration or no filter, with or without locking ports for use with the BacterioScan 216Dx Laser Microbial Growth Monitor.
  • Straight or staggered
  • Sterile or nonsterile
  • Filtration or No Filter
  • 64/Case
Specifications
Sterility Sterile
Type Filtration Multicuvette
Includes 5 μm; Locking Ports
Quantity 256/Cs.
For Use With (Equipment) BacterioScan 216Dx Laser Microbial Growth Monitor
Shape Straight