Learn More
BacterioScan™ 216Dx Microbial Multicuvettes
Catalog No. 22011120
Change view
Click to view available options
Product Type:
Filtration Multicuvette
Multicuvette
Includes:
5 μm; Locking Ports
Non-locking Ports
Sterility:
Non-sterile
Sterile
Description:
Straight
Straight; No Filter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes Sterility Description
22-011-120 Multicuvette Non-locking Ports Non-sterile Straight; No Filter
22-011-105 Filtration Multicuvette 5 μm; Locking Ports Sterile Straight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-011-120 Supplier BacterioScan™ Supplier No. 4DXSAP064
Multicuvettes: Filtration or no filter, with or without locking ports for use with the BacterioScan 216Dx Laser Microbial Growth Monitor.
  • Straight or staggered
  • Sterile or nonsterile
  • Filtration or No Filter
  • 64/Case
Specifications
Product Type Multicuvette
Description Straight; No Filter
Shape Straight
Sterility Non-sterile
Includes Non-locking Ports
Quantity 256/Cs.