Learn More
Lascar Electronics Inc 21CFR USB Vaccine Monitoring Kit
SDP
Catalog No. NC1856623
Encompass_Preferred
This 21CFR-compliant standalone data logger records and displays over 32,000 temperature readings from the supplied thermistor probe. The data logger features an LCD and push-button which allow the user to cycle through the most recent, highest and lowest stored readings.
Catalog No. NC1856623 Supplier Lascar Electronics Inc Supplier No. EL21CFRVAC
This 21CFR-compliant standalone data logger records and displays over 32,000 temperature readings from the supplied thermistor probe. The data logger features an LCD and push-button which allow the user to cycle through the most recent, highest and lowest stored readings.