Learn More
SAI Infusion Technologies 20 Gauge, 0.5" Blunt Needles - Non-sterile, 1000/pk
SDP
Catalog No. NC0099382
Encompass_Preferred
21G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 1000. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC0099382 Supplier SAI Infusion Technologies Supplier No. B2150 1000 BULK
21G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 1000. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.