Learn More
Medchemexpress LLC 21H,23H-porphine, 5,10,15,20-tetrakis(4-methylphenyl) | 14527-51-6 | 97.0% | 670.84 g/mol | C48H38N4 | 5 G
SDP
Catalog No. 5000299302
Encompass_Preferred
5,10,15,20-Tetrakis(p-tolyl)porphyrin is a synthetic porphyrin with a porphyrin core substituted by four p-tolyl groups. It serves as a research reagent and functional material in photonic and catalytic applications, commonly used as a sensitizer in dye-sensitized solar cells and as a catalyst in organic synthesis.
  • High purity (97.0%) suitable for research and synthesis
  • Porphyrin core with p-tolyl substituents enhances photophysical properties
  • Well-defined molecular weight (670.84 g/mol) for stoichiometric calculations
  • Available in small lab-scale quantities for experimental work
  • Useful as a sensitizer in solar cells and as a catalyst in organic reactions
Catalog No. 50-002-99302 Supplier Medchemexpress LLC Supplier No. HYW0351335G
5,10,15,20-Tetrakis(p-tolyl)porphyrin is a synthetic porphyrin with a porphyrin core substituted by four p-tolyl groups. It serves as a research reagent and functional material in photonic and catalytic applications, commonly used as a sensitizer in dye-sensitized solar cells and as a catalyst in organic synthesis.
  • High purity (97.0%) suitable for research and synthesis
  • Porphyrin core with p-tolyl substituents enhances photophysical properties
  • Well-defined molecular weight (670.84 g/mol) for stoichiometric calculations
  • Available in small lab-scale quantities for experimental work
  • Useful as a sensitizer in solar cells and as a catalyst in organic reactions