Learn More
Chemglass Life Sciences 21 x 70 Vials, Amber, 16mL (4 Dram) Capacity, 18-400 Thread with PTFE Lined Solid PTFE Caps, Sold in a LAB-PAC of 144pcs
SDP
Catalog No. 50873524
Encompass_Preferred
21x 70 Vials, Amber, 16mL (4 Dram) Capacity, 18-400 Thread with Solid PTFE Caps, Sold In A LAB-PAC of 144 pcs. Amber sample vials packaged in convenient LAB-PACS of 144 pieces. Made from Type I borosilicated glass. Full height partitions minimize breakage. Solid PTFE lined caps. Open top caps with septa are available under part number CG-4910.
Catalog No. 50-873-524 Supplier Chemglass Life Sciences Supplier No. CG490503
21x 70 Vials, Amber, 16mL (4 Dram) Capacity, 18-400 Thread with Solid PTFE Caps, Sold In A LAB-PAC of 144 pcs. Amber sample vials packaged in convenient LAB-PACS of 144 pieces. Made from Type I borosilicated glass. Full height partitions minimize breakage. Solid PTFE lined caps. Open top caps with septa are available under part number CG-4910.