Learn More
MSC Thomas & Betts 2A-18 22 to 18 AWG Compatible, Noninsulated, Crimp-On Butt Splice Terminal 2 Wire Entries, Copper Contacts, Tin Contact Plating, 0.62" Overall Length, Red
Catalog No. 501297392
Encompass
Thomas & Betts 2A-18 22 to 18 AWG Compatible, Noninsulated, Crimp-On Butt Splice Terminal 2 Wire Entries, Copper Contacts, Tin Contact Plating, 0.62" Overall Length, Red
Catalog No. 50-129-7392 Supplier MSC Supplier No. 54063268
Thomas & Betts 2A-18 22 to 18 AWG Compatible, Noninsulated, Crimp-On Butt Splice Terminal 2 Wire Entries, Copper Contacts, Tin Contact Plating, 0.62" Overall Length, Red