Learn More
MSC 22 3/32X7 1/16 RADIAL SEAL
Catalog No. 5000504335
Encompass
Radial seal air filters prevent airborne contaminants from reaching the combustion chamber, ensuring proper equipment function.
  • Designed as an inner air element
  • Fits inside RS3826
  • Outside diameter 7 1/16 in (179.4 mm)
  • Inside diameter 5 11/16 in (144.5 mm)
  • Length 22 3/32 in (561.2 mm)
  • Suitable for Caterpillar Gen Sets, New Holland, and Volvo equipment
Catalog No. 50-005-04335 Supplier MSC Supplier No. 10949139
Radial seal air filters prevent airborne contaminants from reaching the combustion chamber, ensuring proper equipment function.
  • Designed as an inner air element
  • Fits inside RS3826
  • Outside diameter 7 1/16 in (179.4 mm)
  • Inside diameter 5 11/16 in (144.5 mm)
  • Length 22 3/32 in (561.2 mm)
  • Suitable for Caterpillar Gen Sets, New Holland, and Volvo equipment