Learn More
C&G Containers 22-400 Open Top Screw Cap, PTFE-Faced Silicone Septum, Autoclavable
SDP
Supplier:  C&G Containers CAH221000I04
Encompass_Preferred
22-400 Open Top Screw Cap with PTFE Septum - Designed for reliable sealing in scientific applications, this cap provides convenient access for syringe sampling while preventing leakage and gas exchange. Ideal for volatile solvents and corrosive chemicals, it combines excellent chemical resistance with autoclavable features, differentiating it from standard caps. Its compatibility with various vials and bottles enhances usability across laboratory settings.
Catalog No. NC2738675