Learn More
Medchemexpress LLC HY-D0020 100mg Medchemexpress, 2,2'-Bipyridine CAS:366-18-7 Purity:>98%
SDP
Catalog No. 502082738
Encompass_Preferred
Medchemexpress, HY-D0020 100mg 2,2'-Bipyridine CAS:366-18-7 2,2'-Bipyridine is the unique molecular scaffold of the bioactive natural products represented by caerulomycins (CAEs) and collismycins (COLs). 2,2'-Bipyridine is extensively used as the core structure of many chelating ligands by acting as a bridge in the arrangement of the catalytic center. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2738 Supplier Medchemexpress LLC Supplier No. HYD0020100MG
Medchemexpress, HY-D0020 100mg 2,2'-Bipyridine CAS:366-18-7 2,2'-Bipyridine is the unique molecular scaffold of the bioactive natural products represented by caerulomycins (CAEs) and collismycins (COLs). 2,2'-Bipyridine is extensively used as the core structure of many chelating ligands by acting as a bridge in the arrangement of the catalytic center. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.