Learn More
Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet With Manual-Closing Doors
GREENER_CHOICE
Catalog No. 19822399 Shop All Justrite Safety Group Products
Change view
Click to view available options
Door Style:
Manual-close
Catalog No. Door Style
19-822-399 Manual-close
1 options
Catalog No. 19-822-399 Supplier Justrite™ Supplier No. 892202
  • $3,014.00
    $2,781.65 / Each of 1
    Save $232.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet With Manual-Closing Doors is specially designed for safely storing acids and corrosives in laboratories.
Justrite blue steel Slimline safety cabinets for corrosives have the same quality safety features as the cabinets for flammables. All models come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet. Manual-close door(s) opens to a full 180 degrees and self-latches when pushed closed.
Specifications
Load Bearing Capacity 22 gal.
Description Cold-rolled Steel (18 ga)
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Material Steel (CR)
Door Style Manual-close
Color Blue