Learn More
Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet w/ 3 Shelves and 1 Self-Closing Door
GREENER_CHOICE
Catalog No. 19822400 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
19-822-400 Blue
1 options
Catalog No. 19-822-400 Supplier Justrite™ Supplier No. 892222
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 22 Gallon Sure-Grip™ EX Slimline Corrosives/Acid Steel Safety Cabinet is specially designed for safely storing acids and corrosives in laboratories.
  • Protect workers, reduce fire and explosion risks, and improve productivity by storing corrosives in code-compliant safety cabinets
  • Come with double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet
  • Designed to meet OSHA and NFPA standards, Sure-Grip EX cabinets are constructed of sturdy 18-gauge thick double-wall welded steel
  • High performance, self-latching doors close easily and securely for maximum protection under fire conditions
  • Fail-safe closing mechanism ensures three-point stainless steel bullet latching system works every time
  • For added security, U-Loc handle comes with a cylinder lock and key set or accepts an optional padlock
  • Haz-Alert reflective warning labels are highly visible under fire conditions or during power outages when illuminated with a flashlight
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof bottom sump
  • They adjust on 3 in. centers for versatile storage
  • Durable lead-free epoxy/polyester powder-coat finish provides excellent chemical resistance
  • Under Fume Hood Cabinets are fully compliant and designed for the safe storage and venting of solvents and flammable liquids
  • Ideal in laboratories under fume hoods, benches, or counters, these cabinets are available in four widths ranging from 24 in. (610mm) to 48 in. (1219mm)
  • Single-door cabinets are offered in either left or right-hinge door closures for easy operation where space is at a premium
  • Back wall vents with built-in flame arresters are positioned at lower left and top right for jurisdictions that mandate venting of vapors
Specifications
Description 18-Gauge Steel
No. of Doors 1
No. of Shelves 3
Weight (English) 184 lb
Door Style Self Closing
Load Bearing Capacity 22 gal.
Dimensions (D x W x H) 18 x 23.25 x 65 in.
Color Blue
Material Steel (CR)