Learn More
BRAIN RESEARCH LABORATORIES Cover glass – #2222-1.5D, 22 x 22mm no1.5 thickness (200pc/pk)
Catalog No. NC1949351
Encompass
Cover glass – #2222-1.5D, 22 x 22mm no1.5 thickness (200pc/pk)
Catalog No. NC1949351 Supplier BRAIN RESEARCH LABORATORIES Supplier No. 22221.5D
Cover glass – #2222-1.5D, 22 x 22mm no1.5 thickness (200pc/pk)