Learn More
Hampton Research 22mm x 0.96mm Thick Siliconized square cover slides, ~750 slides
SDP
Catalog No. NC1315573
Encompass
Siliconized glass cover slides allow a droplet to be suspended in a position which provides near optimal conditions for vapor diffusion with the surrounding reservoir solution.
Catalog No. NC1315573 Supplier Hampton Research Supplier No. HR3225
Siliconized glass cover slides allow a droplet to be suspended in a position which provides near optimal conditions for vapor diffusion with the surrounding reservoir solution.