Learn More
AdipoGen 22-NHC . hydrochloride (1 mg)
SDP
Supplier:  AdipoGen AGCR13523M001
Encompass_Preferred
Chemical. CAS: 360068-87-7 or 3915-24-0 (free base). Formula: C26H45NO . HCl. MW: 387.6 . 36.5. Aza-steroid. Hedgehog signaling inhibitor. Competes with 20(S)-OHC for access to the cysteine-rich domain in Smoothened (Smo). For control compound see 22-NHC (inactive isomer) (Prod. No. AG-CR1-3524) http://www.adipogen.com/ag-cr1-3524/22-nhc-inactive-isomer.html .
Catalog No. 50-205-3907