Learn More
AdipoGen 22-NHC (inactive isomer) (5 mg)
SDP
Supplier:  AdipoGen AGCR13524M005
Encompass_Preferred
Chemical. CAS: 40521-33-3. Formula: C26H45NO. MW: 387.6. Inactive control compound for hedgehog signaling inhibitor 22-NHC . HCl (Prod. No. AG-CR1-3523) http://www.adipogen.com/ag-cr1-3523/22-nhc-hydrochloride.html .
Catalog No. 50-205-3910