Learn More
Chemglass Life Sciences PFA Coated Stainless Steel Thermocouple: 1/4" Diameter x 22" Long, Type "T"
SDP
Catalog No. NC0337916
Encompass_Preferred
PFA Coated S.S. Thermocouple: 1/4" Diameter X 22" Long, Type "T"
Catalog No. NC0337916 Supplier Chemglass Life Sciences Supplier No. CG3498122
PFA Coated S.S. Thermocouple: 1/4" Diameter X 22" Long, Type "T"