Learn More
Cayman Chemical 22(S),23(S)-Homobrassinolide, Purity: ≥90%, CAS: 80483-89-2, 10 mg
SDP
Catalog No. 502948087
Encompass_Preferred
A synthetic brassinosteroid; increases the rooting percentage in P. abies cuttings when applied topically at 60 ppm
Catalog No. 50-294-8087 Supplier Cayman Chemical Supplier No. 3288410 MG
A synthetic brassinosteroid; increases the rooting percentage in P. abies cuttings when applied topically at 60 ppm