Learn More
Cayman Chemical 22 S 23 S-HomobrasInolIde 50mg
SDP
Catalog No. 502946943
Encompass_Preferred
A synthetic brassinosteroid; increases the rooting percentage in P. abies cuttings when applied topically at 60 ppm
Catalog No. 50-294-6943 Supplier Cayman Chemical Supplier No. 3288450 MG
A synthetic brassinosteroid; increases the rooting percentage in P. abies cuttings when applied topically at 60 ppm