Learn More
TECHNOTRADE INTERNATIONAL INC Deconex 22 LIQ-X, 6kg
Catalog No. NC0491051
Encompass_Preferred
Liquid, alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material. Rinsable without residues and guarantees therefore consistent and error-free results. With a broad cleaning spectrum and perfect results even at low concentrations. High cleaning performance due to excellent dispersing capabilities. Phosphate- and surfactant-free, no release of phosphates and surfactants into waste water. EDTA-, NTA- and chlorine-free. Rinsing with an acidic deconex cleaner is recommended as the completion of the cleaning process. 6kg
This item is not returnable. View return policy
Catalog No. NC0491051 Supplier TECHNOTRADE INTERNATIONAL INC Supplier No. 519200.00KK5W
Liquid, alkaline cleaning concentrate for automated cleaning of laboratory glassware und reusable laboratory material. Rinsable without residues and guarantees therefore consistent and error-free results. With a broad cleaning spectrum and perfect results even at low concentrations. High cleaning performance due to excellent dispersing capabilities. Phosphate- and surfactant-free, no release of phosphates and surfactants into waste water. EDTA-, NTA- and chlorine-free. Rinsing with an acidic deconex cleaner is recommended as the completion of the cleaning process. 6kg