Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, 19mm, Heavy Duty, for 20000mL Flasks, 39mm Width x 160mm Length, 45/50 minimum joint clearance
SDP
Catalog No. CG208905
Encompass_Preferred
Stirrer Blade, PTFE, 19mm, Heavy Duty, for 22000mL Flasks, 39mm Width x 160mm Length, 45/50 minimum joint clearance. Stirrer blade made of 3/16" thick solid PTFE for use with both Chem-Stir and button style stirring shafts having a 19mm OD.
Catalog No. CG208905 Supplier Chemglass Life Sciences Supplier No. CG208905
Stirrer Blade, PTFE, 19mm, Heavy Duty, for 22000mL Flasks, 39mm Width x 160mm Length, 45/50 minimum joint clearance. Stirrer blade made of 3/16" thick solid PTFE for use with both Chem-Stir and button style stirring shafts having a 19mm OD.