Learn More
Chemglass Life Sciences Glas-col Mantle Tripod Support for Series inMin, Adjustable Height, Mantle Size 22000mL, 489mm ID
SDP
Catalog No. CG1100626
Encompass_Preferred
Glas-col Mantle Tripod Support for Series inMin, Adjustable Height, Mantle Size 22000mL, 489mm ID. Glas-Col tripod support sits on the floor or a bench to hold a Series M mantle ranging in sizes from 3 to 22 liters. Mounting holes are in the foot of each leg for securing to floor or bench. Support is painted gray and shipped unassembled. Adjustable in heights form 24 to 36 in 3 increments.
Catalog No. CG1100626 Supplier Chemglass Life Sciences Supplier No. CG1100626
Glas-col Mantle Tripod Support for Series inMin, Adjustable Height, Mantle Size 22000mL, 489mm ID. Glas-Col tripod support sits on the floor or a bench to hold a Series M mantle ranging in sizes from 3 to 22 liters. Mounting holes are in the foot of each leg for securing to floor or bench. Support is painted gray and shipped unassembled. Adjustable in heights form 24 to 36 in 3 increments.