Learn More
StatLab MasterTech Pentachrome Stain Kit, Russell Movat
SDP
Catalog No. 22021363
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-021-363 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-363 Supplier StatLab Supplier No. KTRMPPT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Pentachrome Stain Kit, Russell Movat - Stains all connective tissue elements, mucins, and nuclei in just over one hour
  • This stain has three primary stains: Verhoeff's Elastic Stain, Alcian Blue, and Crocein Scarlet which produce purple to black elastic fibers and nuclei blue to green mucins, red muscle, and red fibrinoid against a yellow background with collagen yellow collagen counterstained by Alcoholic Saffron Solution
  • Pint Stain Kit yields between 144 to 225 slides and actual slide yields mya vary
  • Pint Kit Consists of - 10% Alcoholic Hematoxylin pint, Universal Iodine Solution pint, 5% Sodium Thiosulfate pint, 1% Alcian Blue pint, 1% Acetic Acid liter, Alcoholic Saffron Solution pint, 10% Ferric Chloride pint, 2% Ferric Chloride pint, 3% Acetic Acid pint, Crocein Scarlet-Acid Fuchsin pint, and 5% Phosphotungstic Acid liter
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • Please review all component labels to ensure correct storage temperature
  • A MasterTech Special Stain Kit
Specifications
Type Russell-Movat Pentachrome Stain
Product Type Special Stain Kit
Quantity 1 pt.