Learn More
StatLab Poly Scientific Weigert's Iron Hematoxylin Solution Set (A and B)

Description
- Often used in Trichrome stains
- Equal parts of A and B are mixed to form the working solution
- Available for purchase in 8 oz, 16 oz, and 32 oz sets
- A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Dark Violet |
| Quantity | 32 oz. |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.