Learn More
ICU Medical Safe-T-Lance™ Safety Lancets
Catalog No. 22024907
Encompass
Change view
Click to view available options
Product Type:
Low Flow
Micro Flow
Pediatric Blade
Color:
Blue
Light Blue
Pink
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
22-024-907 Micro Flow Light Blue
22-024-908 Pediatric Blade Pink
22-024-906 Low Flow Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-024-907 Supplier ICU Medical Supplier No. 10251
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safe-T-Lance™ Safety Lancets with retractable safety technology for capillary blood sampling
  • Retractable safety technology for capillary blood sampling
  • Automatic retraction reduces needlestick injuries and cross-contamination exposure
  • Provides a quick, gentle puncture
  • Needle is shielded before and after use
  • Available in six different sizes for multiple types of applications
  • Lancets are color coded for easy reference
Specifications
Disposable Single-use
Product Type Micro Flow
Color Light Blue
Depth (English) Needle Built-in Control
Needle Gauge 28 ga
Quantity 200/Box