Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Epredia™ Disposable Base Molds
Shop All Epredia Products
                    Click to view available options
                
                
                    
                            
                                Dimensions (L x W):
                            
                        
                        
                            
                                15 x 15 mm
                            
                        
                            
                                24 x 24 mm
                            
                        
                            
                                30 x 24 mm
                            
                        
                            
                                37 x 24 mm
                            
                        
                    Description
- Cleaning associated with metal molds is not required
 - Transparency facilitates tissue orientation
 
Ordering Information:
- Sold in cases of four boxes.
 
Specifications
Specifications
| Product Type | Base Mold | 
| Material | Plastic | 
| Dimensions (L x W) | 15 x 15 mm | 
| Disposable | Single-use | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction