Learn More
Highres Biosolutions Inc 220UL 96 FILTERED NON-STERI
Supplier: Highres Biosolutions Inc 1076350
- Nominal volume: 220 µL
- Volume range: 1-165 µL
- Format: 96-well
- Tip type: Filtered
- Sterility: Non-sterile
- Material: 100% virgin polypropylene, polyethylene, or polystyrene
- Certifications: USP Class-VI certified, RoHS, FDA CFR 21, REACH compliant
- Aerosol protection: true
- Chemical compatibility: Suitable for volatile or corrosive fluids
- Typical applications: Genomics workflows, NGS library preparation, infectious sample assays
- Coefficient of variation (CV): <1.5% at 110 µL and 220 µL test volumes
- Accuracy: <2% at 110 µL and 220 µL test volumes
- Automation compatibility: HighRes Prime automated liquid handlers, third-party automated systems
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.