Learn More
Sigma Aldrich Fine Chemicals Biosciences 224 Trimethylpentane ACS r
Catalog No. 502907062
Encompass
2,2,4-Trimethylpentane (TMP, isooctane), a volatile organic solvent is used to improve the octane rating of combustion fuels. TMP is a primary reference fuel with an arbitrary octane number of 100 as it burns smoothly without resulting in any knocking. It also a component of gasoline. The physical characteristics of the binary mixtures of n-heptane and 2,2,4-trimethylpentane has been studied that helps in understanding the combustion process of mixtures of primary reference fuels.
Catalog No. 50-290-7062 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3605974X4L
2,2,4-Trimethylpentane (TMP, isooctane), a volatile organic solvent is used to improve the octane rating of combustion fuels. TMP is a primary reference fuel with an arbitrary octane number of 100 as it burns smoothly without resulting in any knocking. It also a component of gasoline. The physical characteristics of the binary mixtures of n-heptane and 2,2,4-trimethylpentane has been studied that helps in understanding the combustion process of mixtures of primary reference fuels.