Learn More
Sigma Aldrich Fine Chemicals Biosciences 224 Trimethylpentane suita
Catalog No. 502907966
Encompass
224-Trimethylpentane (TMP Isooctane) is a saturated branched hydrocarbon. It has been reported to be nephrotoxic to male rats. Density and viscosity of TMP at various temperatures (198K - 348K) and pressures (100MPa) has been evaluated. It has been indentified as nephrotoxicity inducing component in unleaded gasoline.
Catalog No. 50-290-7966 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348624X4L
224-Trimethylpentane (TMP Isooctane) is a saturated branched hydrocarbon. It has been reported to be nephrotoxic to male rats. Density and viscosity of TMP at various temperatures (198K - 348K) and pressures (100MPa) has been evaluated. It has been indentified as nephrotoxicity inducing component in unleaded gasoline.