Learn More
eMolecules​ 2247367-07-1 | 4,4,5,5-tetraethyl-2-(4,4,5,5-tetraethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane | Pharmablock366.160 | C20H40B2O4 | 97.000 | CCC1(CC)OB(OC1(CC)CC)B1OC(CC)(CC)C(CC)(CC)O1 | 250g | 867026732
Catalog No. 5000482677 Shop All eMolecules Inc. Products
Encompass_Preferred
4,4,5,5-tetraethyl-2-(4,4,5,5-tetraethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane | Pharmablock | 2247367-07-1366.160 | C20H40B2O4 | 97.000 | CCC1(CC)OB(OC1(CC)CC)B1OC(CC)(CC)C(CC)(CC)O1 | 250g | 867026732
Catalog No. 50-004-82677 Supplier eMolecules​ Supplier No. 867026732
4,4,5,5-tetraethyl-2-(4,4,5,5-tetraethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane | Pharmablock | 2247367-07-1366.160 | C20H40B2O4 | 97.000 | CCC1(CC)OB(OC1(CC)CC)B1OC(CC)(CC)C(CC)(CC)O1 | 250g | 867026732