Learn More
Labcon North America 225mL PerformR Centrifuge Tubes, in Bags
Catalog No. NC1349296
Encompass_Preferred
Tube, Conical centrifuge; Labcon; PerformR; 225mL; With attached flat integral elastomeric seal screw caps, side graduations, and writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 8,000 x G; Freezable to -80C; Autoclavable; Capacity: 225mL; Teal color caps; 2 per bag/24 bags per case; 61mm W. 137mm H; Made in USA
Catalog No. NC1349296 Supplier Labcon North America Supplier No. 30818200009
Tube, Conical centrifuge; Labcon; PerformR; 225mL; With attached flat integral elastomeric seal screw caps, side graduations, and writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 8,000 x G; Freezable to -80C; Autoclavable; Capacity: 225mL; Teal color caps; 2 per bag/24 bags per case; 61mm W. 137mm H; Made in USA