Learn More
Chemglass Life Sciences 225cm² Cell Culture Flask, TC, Plug Seal Cap, Sterile, 5/pk, 25/cs
SDP
Catalog No. 501897654
Encompass_Preferred
Made from high clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase-FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingSolid Plug Seal CapsTissue culture treated cell culture flasks are available in cell growth areas ranging from 25cm¿ to 225cm¿.
Catalog No. 50-189-7654 Supplier Chemglass Life Sciences Supplier No. CGN2405225
Made from high clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase-FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingSolid Plug Seal CapsTissue culture treated cell culture flasks are available in cell growth areas ranging from 25cm¿ to 225cm¿.