Learn More
Alkali Scientific CellPro™ 225cm2 Cell Culture Flask, Vent Cap, TC, Sterile 5/pk, 25/cs
SDP
Catalog No. NC1973729
Encompass_Preferred
CellPro™ Vented TC- Treated Cell Culture Flasks are packaged in zip bags. They are high clarity, 100% virgin polystyrene, stackable, sterilized by E-Beam, and have frosted writing and clear graduations. They have vent caps with breathable 0.22um membrane, clear lot number for batch traceability and are non-pyrogenic, DNase/RNase free.
Catalog No. NC1973729 Supplier Alkali Scientific Supplier No. TVN0225
CellPro™ Vented TC- Treated Cell Culture Flasks are packaged in zip bags. They are high clarity, 100% virgin polystyrene, stackable, sterilized by E-Beam, and have frosted writing and clear graduations. They have vent caps with breathable 0.22um membrane, clear lot number for batch traceability and are non-pyrogenic, DNase/RNase free.