Learn More
NEST SCIENTIFIC INC 225cm2 Cell Culture Flask, Vent Cap, TC, sterile, 25/cs
Catalog No. NC2887672
Encompass_Preferred
Nest Scientific tissue culture flasks are molded from crystal clear USP grade VI virgin polystyrene. They are plasma etched (TC treated) to support adherent cell lines. Feature a vented cap with a USP grade VI 0.22uM PTFE membrane for gas exchange. Flask upper and lower are ultrasonically welded together with no adhesives that may leach. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. This flask is among our best selling items worldwide. ~20% of US brands on US market private label this flask from Nest. cGMP compliant (21 CFR part 820/821). Extensive validation dossiers available upon request.
Catalog No. NC2887672 Supplier NEST SCIENTIFIC INC Supplier No. 721003
Nest Scientific tissue culture flasks are molded from crystal clear USP grade VI virgin polystyrene. They are plasma etched (TC treated) to support adherent cell lines. Feature a vented cap with a USP grade VI 0.22uM PTFE membrane for gas exchange. Flask upper and lower are ultrasonically welded together with no adhesives that may leach. Molded in an ISO 7 cleanroom, sterile (SAL 1 X 10^-6) and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Extensively tested for leachables/extractables per ISO 10993. This flask is among our best selling items worldwide. ~20% of US brands on US market private label this flask from Nest. cGMP compliant (21 CFR part 820/821). Extensive validation dossiers available upon request.