Learn More
Apex Aseptic Extra-Safe™ Lab Jackets, Ceil Blue
Catalog No. 22770022
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-770-022 X-Large
22-770-019 Small
22-770-020 Medium
22-770-021 Large
22-770-023 2X-Large
22-770-024 3X-Large
22-770-025 4X-Large
22-770-026 5X-Large
22-770-018 X-Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-770-022 Supplier Apex Aseptic Supplier No. 3630CBXL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Static-free unisex lab coat is made with a 3-layer SMS material to provide protection and comfort in a variety of environments.
  • Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabric
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Hip length
  • Autoclavable and washable up to five times
  • Latex-free
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color Ceil Blue
Closure Type Snaps
Cuff Style Knit
Disposable Limited-use
Sterility Non-sterile
Resistance Fluids
Style Hip-length
No. of Pockets 3
Collar Type Knit
Latex Free Yes
Model 3630CB
Thickness 3-ply
Autoclavable Not Autoclavable