Learn More
SAI Infusion Technologies Stainless Steel Catheter Plugs - 22G, 1cm, 100/pk
SDP
Catalog No. NC9995641
Encompass_Preferred
SAI's Stainless Steel Catheter Plugs safely seal catheters when not in use. Burr-free stainless steel prevents damage to the catheter during insertion and removal. Catheter pins are solid. To connect two catheters instead, use hollow steel catheter connectors (CC-22). Non-sterile, 22G, 1cm, 100/pk. Product is for laboratory animal research only.
Catalog No. NC9995641 Supplier SAI Infusion Technologies Supplier No. CP22
SAI's Stainless Steel Catheter Plugs safely seal catheters when not in use. Burr-free stainless steel prevents damage to the catheter during insertion and removal. Catheter pins are solid. To connect two catheters instead, use hollow steel catheter connectors (CC-22). Non-sterile, 22G, 1cm, 100/pk. Product is for laboratory animal research only.