Learn More
SAI Infusion Technologies Stainless Steel Catheter Plugs - 22G, 1cm, 25/pk
SDP
Catalog No. NC1486077
Encompass_Preferred
SAI's Stainless Steel Catheter Plugs safely seal catheters when not in use. Burr-free stainless steel prevents damage to the catheter during insertion and removal. Catheter pins are solid. To connect two catheters instead, use hollow steel catheter connectors (CC-22). Non-sterile, 22G, 1cm, 25/pk. Product is for laboratory animal research only.
This item is not returnable. View return policy
Catalog No. NC1486077 Supplier SAI Infusion Technologies Supplier No. CP2225
SAI's Stainless Steel Catheter Plugs safely seal catheters when not in use. Burr-free stainless steel prevents damage to the catheter during insertion and removal. Catheter pins are solid. To connect two catheters instead, use hollow steel catheter connectors (CC-22). Non-sterile, 22G, 1cm, 25/pk. Product is for laboratory animal research only.