Learn More
SAI Infusion Technologies 22 Gauge, 0.5" Blunt Needles - Non-sterile, 100/pk
SDP
Catalog No. NC1362045
Encompass
22G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC1362045 Supplier SAI Infusion Technologies Supplier No. B2250 100 BULK
22G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.